missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconyl chloride octahydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White to Yellow |
Physical Form | Crystalline Powder |
Quantity | 100 g |
Assay Percent Range | 97.5 to 102.5% (Argentometry) |
Linear Formula | ZrOCl2·8H2O |
Merck Index | 15, 10384 |
Solubility Information | (10% in water) Clear to slightly hazy solution |
Formula Weight | 322.25 |
Percent Purity | 98% |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- Zirconyl chloride octahydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Zirconyl chloride octahydrate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction