missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ZNRF4 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Marca: Abnova™ H00148066-T01.100uL
Detalles adicionales : Weight : 0.00010kg
Especificaciones
Western Blot | |
ZNRF4 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 47.3kDa | |
Transient overexpression cell lysate was tested with anti-ZNRF4 antibody (Mfr. No. H00148066-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
NM_181710.2 | |
pCMV-ZNRF4 full-length | |
100 μL | |
Human |
Seguridad y manipulación
Nombre del producto
Palabra de advertencia
Clasificación
Indicaciones de peligro
Consejos de prudencia
Otros peligros
- 100UL ZNRF4 293T Cell Transient Overexpression Lys
Palabra de advertencia
- Warning
Clasificación
- Skin sensitiser Category 1
Indicaciones de peligro
- H317-May cause an allergic skin reaction.
Consejos de prudencia
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Otros peligros
- MIXTURE LIST-Contains : Mercaptoethanol