Learn More
Specac™ ZnSe Sealed Heatable Liquid Static Cell
Brand: Specac™
Code : Z3
Additional Details : Weight : 0.01000kg
Description
This heatable liquid cell is designed for high sensitivity ATR measurements with FTIR or dispersive instruments. This is a sealed cell with ZnSe windows with a choice of different pathlengths available ranging from 0.025mm up to 1.0mm.Specifications
Pathlengths available ranging from 0.025 mm up to 1.0 mm |