Learn More
Zofenopril calcium, MedChemExpress
MedChemExpress Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.
Brand:  Medchem Express HY-B0655
Encompass_Preferred
 View more versions of this product
Product Code. 16411038
  • £85.91 / 5mg
Explore more special offers
Description
Description
Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.
Specifications
Specifications
Zofenopril calcium
White
C22H22Ca0.5NO4S2
SQ26991
O=C(N(C[C@@H](SC1=CC=CC=C1)C2)[C@@H]2C([O-])=O)[C@H](C)CSC(C3=CC=CC=C3)=O.[0.5Ca2+]
448.58
Research
Neuroscience-Neurodegeneration
81938-43-4
Research
5 mg
DMSO : 2 mg/mL (4.46 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
448.58
99.56%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Zofenopril calcium, MedChemExpress