Learn More
Zolbetuximab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand:  Invitrogen MA542158
286.30 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Product Code. 17708783
  • £409.00 / 100µg
Explore more special offers
Description
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Zolbetuximab is under investigation in clinical trial NCT01630083 (Efficacy and Safety of IMAB362 in Combination With the EOX Regimen for CLDN18. 2-positive Gastric Cancer).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zolbetuximab
Recombinant Monoclonal
Unconjugated
claudiximab; GC-182; IMAB362; IMAB-362
Human CLDN18
100 μg
Primary
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA
3.3 mg/mL
PBS with no preservative; pH 7.4
Human
Protein A
RUO
Chemical
Antibody
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Zolbetuximab Recombinant Human Monoclonal Antibody, Invitrogen™ > 100 μg; Unconjugated