Learn More
Invitrogen™ ZOOM™ IPGRunner™ Cassettes and Trays
Convenient, disposable trays
Brand: Invitrogen™ ZM0008
Code : 50
Additional Details : Weight : 0.54000kg
Includes: Box of 10 cassettes and a pack of 10 trays
Description
ZOOM Equilibration Trays are designed for use with the ZOOM IPGRunner Cassettes to perform equilibration of ZOOM Strips after first-dimension isoelectric focusing and prior to second-dimension (2D) SDS-PAGE.
- Simplify the equilibration and incubation process in 2D electrophoresis
- Savings in time and labor
- Optimal equilibration of samples
- Minimal risk of sample contamination and damage to IPG strips
2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Specifications
The ZOOM™ Equilibration Trays are supplied in a pack of 10. The combo kit of ZOOM™Cassettes and ZOOM™ Equilibration Trays includes a box of 10 cassettes and a pack of 10 trays. Store at room temperature. Guaranteed stable for one year when properly stored. | |
ZOOM IPGRunner System | |
IPGRunner, ZOOM |
For Research Use Only. Not for use in diagnostic procedures.