Learn More
Invitrogen™ ZOOM™ IPGRunner™ Mini-cell
Oil-free, trouble-free 2D electrophoresis in a minigel format
Brand: Invitrogen™ ZM0001
Code : 50
Additional Details : Weight : 0.56000kg
Includes: ZOOM IPGRunner Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini-Cell chamber
Description
The ZOOM IPGRunner System consists of three main components. The ZOOM IPGRunner Mini-Cell includes a high-voltage electrophoresis core and lid assembly that fits a mini-cell chamber identical to the XCell SureLock™ mini-cell. The mini-cell holds two ZOOM IPGRunner Cassettes. The ZOOM IPGRunner Cassettes (available separately) are provided ready to use and self-contained, eliminating oil overlays for convenient sample rehydration and IEF. Each cassette holds up to six ZOOM Strips. The ZOOM Strips (available separately) are 7cm long and contain a thin layer of polyacrylamide gel that includes a fixed pH gradient. Each ZOOM Strip is clearly labeled with a unique identifying number, pH range, and orientation marks. ZOOM Strips are supplied attached to a tri-fold card for easy access and removal.
- The ZOOM IPGRunner System typically completes first dimension separation isoelectric focusing (IEF) in less than three hours
- The minigel design of the system is easy to handle, eliminates mineral oil overlays, and allows processing of up to 12 samples at once for high-throughput usage
Quick and Simple Set-up Procedure
- Easily load samples and set up the ZOOM IPGRunner System for IEF
- Once IEF is complete, the ZOOM IPGRunner Mini-Cell can be used with NuPAGE™ and Novex™ ZOOM™ gels to complete SDS-PAGE analysis in 40 minutes
2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Specifications
The ZOOM IPGRunner Mini-Cell includes the ZOOM IPGRunner Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini- Cell chamber. | |
ZOOM™ IPG Strips | |
IPGRunner, ZOOM |
XCell SureLock™ Mini, ZOOM™ IPGRunner™ Mini | |
7 cm Strips |
For Research Use Only. Not for use in diagnostic procedures.