missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZOOM™ IPGRunner™ Retrofit Kit

Compatible with XCell SureLock Mini-Cell only

Brand:  Invitrogen™ ZM0004

596.40 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10063592

  • £840.00 / 1 set
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: ZOOM IPGRunner Buffer Core and Lid
This item is not returnable. View return policy

Description

Description

The ZOOM IPGRunner System offers oil-free, trouble-free 2D electrophoresis in a minigel format. The system consists of three main Components. The ZOOM IPGRunner Mini-Cell includes a high-voltage electrophoresis core and lid assembly that fits a mini-cell chamber identical to the XCell SureLock™ mini-cell and holds two ZOOM IPGRunner Cassettes. The ZOOM IPGRunner Cassettes are provided ready-to-use and self-contained, eliminating oil overlays for convenient sample rehydration and IEF. Each cassette holds up to six ZOOM Strips. The ZOOM Strips are 7cm long and contain a thin layer of polyacrylamide gel that includes a fixed pH gradient. Each ZOOM Strip is clearly labeled with a unique identifying number, pH range, and orientation marks. ZOOM Strips are supplied attached to a tri-fold card for easy access and removal.

  • First dimension separation isoelectric focusing (IEF) is complete in less than three hours
  • The mini-gel design of the system is easy to handle, eliminates mineral oil overlays, and allows processing of up to 12 samples at once for high-throughput usage

Quick and Simple Set-up Procedure

  • Easy and fast to load samples and set up the ZOOM IPGRunner System for IEF
  • Once IEF is complete, the ZOOM IPGRunner Mini-Cell can be used with NuPAGE™ and Novex™ ZOOM™ gels to complete SDS-PAGE analysis in 40 minutes

2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY
Specifications

Specifications

The ZOOM™ IPGRunner™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini- Cell chamber. The ZOOM™ IPGRunner™ Combo Kit includes the ZOOM™ IPGRunner™ Mini-Cell, electrode wicks, sealing tape, 10 ZOOM™ IPGRunner™ Cassettes, and 12 ZOOM™ Strips, pH 3-10 NL. The ZOOM™ IPGRunner™ Retrofit Kit for XCell SureLock™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid. The ZOOM™ IPGRunner™ Cassettes include electrode wicks and sealing tape.
ZOOM™ IPG Strips
IPGRunner, ZOOM
XCell SureLock™ Mini, ZOOM™ IPGRunner™ Mini
7 cm Strips
1 kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.