Learn More
Thermo Scientific™ Zopiclone
Brand:  Thermo Scientific™ 467230010
 View more versions of this product
Product Code. 17221302
  • £320.00 / 1g
Explore more special offers
Specifications
Zopiclone
1 g
10245
C17 H17 Cl N6 O3
Powder
43200-80-2
soluble in acetone, chloroform, DMSO
173°C to 175°C
388.81
White to Light-yellow