Learn More
Thermo Scientific™ Zopiclone
Brand:  Thermo Scientific™ 467232500
 View more versions of this product
Product Code. 17231302
  • £101.51 / 250mg
Explore more special offers
Specifications
Zopiclone
250 mg
10245
C17 H17 Cl N6 O3
Powder
43200-80-2
soluble in acetone, chloroform, DMSO
173°C to 175°C
388.81
White to Light-yellow