missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Zopiclone
£101.51 - £320.00
Specifications
Chemical Name or Material | Zopiclone |
---|---|
CAS | 43200-80-2 |
Solubility | soluble in acetone, chloroform, DMSO |
Merck Index | 10245 |
Melting Point | 173°C to 175°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17221302
|
Thermo Scientific™
467230010 |
1 g |
£320.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17231302
|
Thermo Scientific™
467232500 |
250 mg |
£101.51
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Zopiclone | |
soluble in acetone, chloroform, DMSO | |
173°C to 175°C | |
388.81 | |
White to Light-yellow |
43200-80-2 | |
10245 | |
C17 H17 Cl N6 O3 | |
Powder |
Safety and Handling
GHS08: Health hazard
GHS07: Harmful
missing translation for 'einecsNumber' : 256-138-9
Recommended Storage : Store at -20°C