missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Zymolyase™
For lysing the cell walls of viable yeast cells and preparing yeast spheroblasts
£891.00 - £1420.00
Specifications
pH | 5 to 10 |
---|---|
For Use With (Application) | Used to lyse living yeast cell walls to produce protoplast or spheroplast of various strains of Yeast cells |
Content And Storage | 2°C to 8°C |
Source | Arthrobacter luteus |
Form | Lyophilized Powder |
Product Code | Brand | Concentration | Product Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Product Type | Quantity | Price | Quantity & Availability | |||
11473556
|
MP Biomedicals™
08320932 |
100,000 U/mg | Zymolyase 100T | 250 mg |
£1420.00
0.25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12760228
|
MP Biomedicals™
08320921 |
20,000 U/mg | Zymolyase 20T | 1 g |
£891.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Highly purified preparation from Arthrobacter luteus
- Efficiently hydrolyzes poly (β-1,3 glucose)-like yeast cell-wall glucan
- One unit is defined as the amount of activity causing a decrease in A800 of 30%
- Available with lytic activity of 20,000U/g (Zymolyase 20T) or 100,000U/g (Zymolyase 100T)
Specifications
5 to 10 | |
2°C to 8°C | |
Lyophilized Powder |
Used to lyse living yeast cell walls to produce protoplast or spheroplast of various strains of Yeast cells | |
Arthrobacter luteus |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Zymolyase 20 T
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Yeast Lytic Enzyme