man

Test

CAS RN 10024-93-8

ClClClNd

CAS RN 10024-93-8

IUPAC Name: neodymium(3+) trichloride
Synonyms: neodymium(III) chloride
Molecular Weight (g/mol): 250.59
Molecular Formula: Cl3Nd
InChi Key: ATINCSYRHURBSP-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Nd+3]
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (1)