man

Test

CAS RN 106-91-2

H3CCH2OOO

CAS RN 106-91-2

IUPAC Name: (oxiran-2-yl)methyl 2-methylprop-2-enoate
Synonyms: glycidyl methacrylate
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: VOZRXNHHFUQHIL-UHFFFAOYNA-N
SMILES: CC(=C)C(=O)OCC1CO1
Boiling Point 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)