man

Test

CAS RN 1071-76-7

ZrCH3HO

CAS RN 1071-76-7

IUPAC Name: butan-1-ol zirconium
Synonyms: butanol zirconium
Molecular Weight (g/mol): 165.35
Molecular Formula: C4H10OZr
InChi Key: SKFIUGUKJUULEM-UHFFFAOYSA-N
SMILES: [Zr].CCCCO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results