missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium n-butoxide, 80% w/w in 1-butanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from oxidizing agents, acids, bases, halogens, alkali metals, water/ moisture.
Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from oxidizing agents, acids, bases, halogens, alkali metals, water/ moisture.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 250 g |
Linear Formula | [CH3(CH2)3O]4Zr |
UN Number | UN1120 |
Beilstein | 4132649 |
Flash Point | 38°C (100°F) |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 383.68 |
Concentration or Composition (by Analyte or Components) | 80% w/w in 1-butanol |
Odor | Alcohol-like |
Refractive Index | 1.466 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'