CAS RN 108-10-1
CAS RN 108-10-1
4-Methyl-2-pentanone, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, 99%, Thermo Scientific Chemicals
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methylpentan-2-One, Extra Pure, SLR, Fisher Chemical™
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: 8938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, ACS, 98.5+%, Thermo Scientific Chemicals
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, 99.5%, spectrophotometric grade, Thermo Scientific Chemicals
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, HPLC Grade, 99+%, Thermo Scientific Chemicals
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methylpentan-2-One, Certified AR for Analysis, Fisher Chemical™
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: 8938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, 98.5+%, ACS reagent, Thermo Scientific Chemicals
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, 99+%, electronic use grade, residue free, Thermo Scientific Chemicals
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, BAKER ANALYZED™ ACS, J.T. Baker™
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: Methyl Isobutyl Ketone , Isobutyl Methyl Ketone , Methyl iso-Butyl Ketone IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(C)=O
4-Methyl-2-pentanone, Honeywell
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, puriss. p.a., ACS Reagent, ≥99%, Honeywell Riedel-de Haën™
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methylpentan-2-One, for AAS, Fisher Chemical™
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: 8938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
- 1
- 2