missing translation for 'onlineSavingsMsg'
Learn More
4-Methyl-2-pentanone, 99.5%, for analysis, Thermo Scientific Chemicals Product Code.: 10266833

4-Methyl-2-pentanone, 99.5%, for analysis, Thermo Scientific Chemicals

Product Code. 10266833
2.5 L, Glass bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
25 L
Packaging:
Glass bottle
Metal drum
Plastic drum
This item is not returnable. View return policy

Product Code. 10266833

Brand: Thermo Scientific Acros 255660025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketonShow MoreShow Less
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Acidity 0.0004 meq/g max.
CAS Min % 99.4
CAS Max % 100.0
Melting Point -84.0°C
Density 0.8000g/mL
Boiling Point 117.4°C
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2COCH3
Refractive Index 1.3950 to 1.3970
Quantity 2.5 L
Beilstein 01,691
Residue after Evaporation 0.005% max.
Merck Index 15,5253
Specific Gravity 0.8
Solubility Information Solubility in water: 17g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,methylene chloride and acetone
Formula Weight 100.16
Percent Purity 99.5%
Grade Analysis
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467700 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number255665000,255660250,255660100,255660010,255660025
Lot Number2410297 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number255665000,255660250,255660100,255660010,255660025
Lot NumberTESTA Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number255665000,255660250,255660100,255660010,255660025
Lot Number2349944 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number255665000,255660250,255660100,255660010,255660025
Lot Number2183926 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number255665000,255660250,255660100,255660010,255660025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methyl-2-pentanone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methyl-2-pentanone, 99.5%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.