CAS RN 172895-15-7
CAS RN 172895-15-7
IUPAC Name:
1-[(3,4-dimethoxyphenyl)methyl]-6-methyl-1H,2H,3H,4H,9H-pyrido[3,4-b]indole hydrochloride
Synonyms:
1-[(3,4-dimethoxyphenyl)methyl]-6-methyl-1H,2H,3H,4H,9H-pyrido[3,4-b]indole hydrochloride
Molecular Weight (g/mol):
372.89
Molecular Formula:
C21H25ClN2O2
InChi Key:
BKAZOTIBKRWLQA-UHFFFAOYNA-N
SMILES:
Cl.COC1=C(OC)C=C(CC2NCCC3=C2NC2=CC=C(C)C=C32)C=C1
1
–
1
of
1
results
1
LY 272015 hydrochloride, Tocris Bioscience™
CAS: 172895-15-7 Molecular Formula: C21H25ClN2O2 Molecular Weight (g/mol): 372.893 InChI Key: BKAZOTIBKRWLQA-UHFFFAOYSA-N Synonym: ly 272015 hydrochloride,1-3,4-dimethoxybenzyl-6-methyl-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole hcl,1-3,4-dimethoxyphenyl methy-2,3,4,9-tetrahydro-6-methyl-1h-pyrido 3,4-b indole hydrochloride,1-3,4-dimethoxyphenyl methyl-6-methyl-1h,2h,3h,4h,9h-pyrido 3,4-b indole hydrochloride PubChem CID: 9929423 IUPAC Name: 1-[(3,4-dimethoxyphenyl)methyl]-6-methyl-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole;hydrochloride SMILES: CC1=CC2=C(C=C1)NC3=C2CCNC3CC4=CC(=C(C=C4)OC)OC.Cl