accessibility menu, dialog, popup

Test

CAS RN 4906-25-6

H3COOOCH3

CAS RN 4906-25-6

IUPAC Name: methyl 4-butoxybenzoate
Synonyms: methyl 4-butoxybenzoate
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: PUHCYTHGLBCWEA-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)C(=O)OC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results