accessibility menu, dialog, popup

Test

CAS RN 733717-87-8

H3COHNOHOCH3

CAS RN 733717-87-8

IUPAC Name: 2-(5,5-dimethylmorpholin-2-yl)acetic acid
Synonyms: (5,5-dimethylmorpholin-2-yl)acetic acid
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: SEYCKMQSPUVYEF-UHFFFAOYNA-N
SMILES: CC1(C)COC(CC(O)=O)CN1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results