man

Test

CAS RN 883031-03-6

CH3NNOONHFFF

CAS RN 883031-03-6

IUPAC Name: 2-methyl-1-[3-(morpholin-4-yl)propyl]-5-phenyl-N-[3-(trifluoromethyl)phenyl]-1H-pyrrole-3-carboxamide
Synonyms: 2-methyl-1-[3-(morpholin-4-yl)propyl]-5-phenyl-N-[3-(trifluoromethyl)phenyl]pyrrole-3-carboxamide
Molecular Weight (g/mol): 471.52
Molecular Formula: C26H28F3N3O2
InChi Key: NCZYSQOTAYFTNM-UHFFFAOYSA-N
SMILES: CC1=C(C=C(N1CCCN1CCOCC1)C1=CC=CC=C1)C(=O)NC1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results