CAS RN 904696-60-2
CAS RN 904696-60-2
IUPAC Name:
lithium(1+) 2-[(morpholin-4-yl)methyl]benzoate
Synonyms:
lithium(1+) 2-(morpholin-4-ylmethyl)benzoate
Molecular Weight (g/mol):
227.19
Molecular Formula:
C12H14LiNO3
InChi Key:
PKLIDBHLNQBFTO-UHFFFAOYSA-M
SMILES:
[Li+].[O-]C(=O)C1=CC=CC=C1CN1CCOCC1