Code:
29
Diethyl phenylmalonate, 98%, Thermo Scientific Chemicals
Additional Details:
CAS Number:
83-13-6
Weight:
0.27250kg
Quantity | 250 mL |
---|---|
Packaging | Glass bottle |
Disclaimers:
RUO – Research Use Only
Chemical Identifiers
CAS | 83-13-6 |
---|---|
MDL Number | MFCD00009144 |
InChI Key | FGYDHYCFHBSNPE-UHFFFAOYSA-N |
Synonym | diethyl phenylmalonate,diethyl 2-phenylmalonate,phenylmalonic acid diethyl ester,propanedioic acid, phenyl-, diethyl ester,diethylphenylmalonate,1,3-diethyl 2-phenylpropanedioate,malonic acid, phenyl-, diethyl ester,propanedioic acid, 2-phenyl-, 1,3-diethyl ester,diethyl 2-phenylpropane-1,3-dioate,diethyl-phenylmalonate |
PubChem CID | 66514 |
IUPAC Name | diethyl 2-phenylpropanedioate |
SMILES | CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC |
Specification
Chemical Name or Material | Diethyl phenylmalonate |
---|---|
CAS | 83-13-6 |
Melting Point | 16.0°C to 17.0°C |
Color | Colorless to Yellow |
Density | 1.0900g/mL |
Boiling Point | 170.0°C to 172.0°C (14.0 mmHg) |
Flash Point | >112°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C13H16O4 |
Refractive Index | 1.4903 to 1.4923 |
Linear Formula | C2H5O2CCH(C6H5)CO2C2H5 |
MDL Number | MFCD00009144 |
Quantity | 250 mL |
Beilstein | 09, 854 |
Specific Gravity | 1.09 |
Synonym | diethyl phenylmalonate,diethyl 2-phenylmalonate,phenylmalonic acid diethyl ester,propanedioic acid, phenyl-, diethyl ester,diethylphenylmalonate,1,3-diethyl 2-phenylpropanedioate,malonic acid, phenyl-, diethyl ester,propanedioic acid, 2-phenyl-, 1,3-diethyl ester,diethyl 2-phenylpropane-1,3-dioate,diethyl-phenylmalonate |
Solubility Information | Solubility in water: immiscible |
InChI Key | FGYDHYCFHBSNPE-UHFFFAOYSA-N |
SMILES | CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC |
IUPAC Name | diethyl 2-phenylpropanedioate |
Molecular Weight (g/mol) | 236.27 |
PubChem CID | 66514 |
Formula Weight | 236.27 |
Percent Purity | 98% |
Physical Form | Liquid After Melting |