missing translation for 'onlineSavingsMsg'
Learn More
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis Product Code.: 10245231

1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis

Product Code. 10245231
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

Product Code. 10245231

Brand: Thermo Scientific Acros 354861000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

CAS: 920-66-1 | C3H2F6O | 168.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 920-66-1
CAS Min % 99.75
CAS Max % 100.0
Melting Point -4.0°C
Density 1.5960g/mL
Boiling Point 59.0°C
Flash Point >100°C
Assay Percent Range 99.8% min. (GC)
Packaging Glass bottle
Molecular Formula C3H2F6O
Linear Formula (CF3)2CHOH
Refractive Index 1.2740 to 1.2770
MDL Number MFCD00011651
Quantity 100 mL
Residue after Evaporation 0.0005% max.
Specific Gravity 1.596
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
Solubility Information Solubility in water: 1000g/L (25°C)
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
SMILES C(C(F)(F)F)(C(F)(F)F)O
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular Weight (g/mol) 168.04
PubChem CID 13529
ChEBI CHEBI:63104
Viscosity 1.65 mPa.s (20°C)
Formula Weight 168.04
Percent Purity 99.8%
Grade For peptide synthesis
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466188 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number354861000
Lot NumberA0462484 Certificate TypeCertificate of Analysis Date06/06/2024 Catalog Number354861000
Lot NumberA0458853 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog Number354861000
Lot NumberA0454827 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number354861000
Lot NumberA0451921 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number354861000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1,1,3,3,3-Hexafluoro-2-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.