missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropanol, >99%, MP Biomedicals™

1,1,1,3,3,3-Hexafluoro-Isopropanol is a speciality solvent effective for solubilizing a wide range of polymers, including those that are not soluble in the most common organic solvents.

£83.70 - £197.00

Chemical Identifiers

CAS 29463-77-2
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.038
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
PubChem CID 13529
ChEBI CHEBI:63104
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11423801
View Documents
MP Biomedicals
0215124525
25 mL
£83.70
25mL
Please to purchase this item. Need a web account? Register with us today!
11433801
View Documents
MP Biomedicals
0215124580
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • 1,1,1,3,3,3-Hexafluoro-Isopropanol is a speciality solvent effective for solubilizing a wide range of polymers, including those that are not soluble in the most common organic solvents, such as:
    • Polyamides, polyacrylonitriles, polyacetals, polyesters (e.g. polyglycolide), and polyketones.
  • It has also found use in biochemistry to solubilize peptides and to monomerize β-sheet protein aggregates.
  • It can be used as an acid in volatile buffers for ion pair HPLC - mass spectrometry of nucleic acids.
  • 1,1,1,3,3,3-Hexafluoro-2-propanol was used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials.
Specifications

Chemical Identifiers

29463-77-2
168.038
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
CHEBI:63104
C(C(F)(F)F)(C(F)(F)F)O
C3H2F6O
BYEAHWXPCBROCE-UHFFFAOYSA-N
13529
1,1,1,3,3,3-hexafluoropropan-2-ol

Specifications

29463-77-2
1.62 g/mL
C3H2F6O
BYEAHWXPCBROCE-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoropropan-2-ol
13529
168.04
Liquid
Colorless
≥99%
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
C(C(F)(F)F)(C(F)(F)F)O
168.038
CHEBI:63104
0.9969
1,1,1,3,3,3-Hexafluoro-isopropanol, For GC analysis
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature (15°C to 30°C).

Product Identifier
  • 1,1,1,3,3,3-hexafluoropropan-2-ol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 1,1,1,3,3,3-hexafluoropropan-2-ol

SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.