missing translation for 'onlineSavingsMsg'
Learn More

1-(1,3-Benzothiazol-2-yl)-2-bromo-1-ethanone, 90+%, Thermo Scientific™

£47.20 - £610.00

Chemical Identifiers

CAS 54223-20-0
Molecular Formula C9H6BrNOS
Molecular Weight (g/mol) 256.12
MDL Number MFCD03659698
InChI Key AYWGYNKWZWBMSV-UHFFFAOYSA-N
Synonym 1-1,3-benzothiazol-2-yl-2-bromo-1-ethanone, 1-benzo d thiazol-2-yl-2-bromoethanone, 1-1,3-benzothiazol-2-yl-2-bromoethanone, 2-bromoacetyl benzothiazole, 1-benzothiazol-2-yl-2-bromoethanone, 1-benzothiazol-2-yl-2-bromo-ethanone, 1-1,3-benzothiazol-2-yl-2-bromoethan-1-one, pubchem23327, 2-bromoacetyl-1,3-benzothiazole
PubChem CID 2776256
SMILES BrCC(=O)C1=NC2=CC=CC=C2S1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10190582
View Documents Promotion Details
Thermo Scientific Maybridge
CC06005CB
250 mg
£47.20
250mg
Please to purchase this item. Need a web account? Register with us today!
10532852
View Documents Promotion Details
Thermo Scientific Maybridge
CC06005DA
1 g
£93.20
1g
Please to purchase this item. Need a web account? Register with us today!
10555144
View Documents Promotion Details
Thermo Scientific Maybridge
CC06005EA
10 g
£610.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

54223-20-0
256.12
AYWGYNKWZWBMSV-UHFFFAOYSA-N
2776256
C9H6BrNOS
MFCD03659698
1-1,3-benzothiazol-2-yl-2-bromo-1-ethanone, 1-benzo d thiazol-2-yl-2-bromoethanone, 1-1,3-benzothiazol-2-yl-2-bromoethanone, 2-bromoacetyl benzothiazole, 1-benzothiazol-2-yl-2-bromoethanone, 1-benzothiazol-2-yl-2-bromo-ethanone, 1-1,3-benzothiazol-2-yl-2-bromoethan-1-one, pubchem23327, 2-bromoacetyl-1,3-benzothiazole
BrCC(=O)C1=NC2=CC=CC=C2S1

Specifications

54223-20-0
C9H6BrNOS
1-1,3-benzothiazol-2-yl-2-bromo-1-ethanone, 1-benzo d thiazol-2-yl-2-bromoethanone, 1-1,3-benzothiazol-2-yl-2-bromoethanone, 2-bromoacetyl benzothiazole, 1-benzothiazol-2-yl-2-bromoethanone, 1-benzothiazol-2-yl-2-bromo-ethanone, 1-1,3-benzothiazol-2-yl-2-bromoethan-1-one, pubchem23327, 2-bromoacetyl-1,3-benzothiazole
BrCC(=O)C1=NC2=CC=CC=C2S1
256.12
256.12
1-(1, 3-Benzothiazol-2-yl)-2-bromo-1-ethanone
90%
MFCD03659698
AYWGYNKWZWBMSV-UHFFFAOYSA-N
1-(1,3-benzothiazol-2-yl)-2-bromoethanone
2776256
90%
Videos
SDS
Documents

Documents

Product Certifications