Learn More
1-(1-Adamantyl)ethylamine hydrochloride, 98%, Thermo Scientific Chemicals
£53.40 - £219.00
Chemical Identifiers
CAS | 1501-84-4 |
---|---|
Molecular Formula | C12H22ClN |
Molecular Weight (g/mol) | 215.77 |
MDL Number | MFCD00072023 |
InChI Key | OZBDFBJXRJWNAV-UHFFFAOYNA-N |
Synonym | rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan |
PubChem CID | 15165 |
ChEBI | CHEBI:8865 |
IUPAC Name | 1-(1-adamantyl)ethanamine;hydrochloride |
SMILES | [H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15410138
|
Thermo Scientific Alfa Aesar
H53377.06 |
5 g |
£53.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15420138
|
Thermo Scientific Alfa Aesar
H53377.14 |
25 g |
£219.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).
Solubility
Freely soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Chemical Identifiers
1501-84-4 | |
215.77 | |
OZBDFBJXRJWNAV-UHFFFAOYNA-N | |
15165 | |
1-(1-adamantyl)ethanamine;hydrochloride |
C12H22ClN | |
MFCD00072023 | |
rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan | |
CHEBI:8865 | |
[H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2 |
Specifications
1501-84-4 | |
C12H22ClN | |
148224 | |
Freely soluble in water. | |
[H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2 | |
215.77 | |
CHEBI:8865 | |
98% |
375°C | |
MFCD00072023 | |
rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan | |
OZBDFBJXRJWNAV-UHFFFAOYNA-N | |
1-(1-adamantyl)ethanamine;hydrochloride | |
15165 | |
215.77 | |
1-(1-Adamantyl)ethylamine hydrochloride |
Safety and Handling
Harmful if swallowed
Acute toxicity (category 4)
missing translation for 'rtecsNumber' : AU4717000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only