missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(1-Benzofuran-3-yl)-2-bromo-1-ethanone, 95+%, Thermo Scientific™
£24.20 - £200.00
Chemical Identifiers
CAS | 187657-92-7 |
---|---|
Molecular Formula | C10H7BrO2 |
Molecular Weight (g/mol) | 239.07 |
MDL Number | MFCD03407369 |
InChI Key | WVHFTONHSRLBGL-UHFFFAOYSA-N |
Synonym | 1-benzofuran-3-yl-2-bromoethanone, 1-1-benzofuran-3-yl-2-bromo-1-ethanone, 1-1-benzofuran-3-yl-2-bromoethanone, 1-benzofuran-3-yl-2-bromoethan-1-one, 1-1-benzofuran-3-yl-2-bromoethan-1-one, ethanone,1-3-benzofuranyl-2-bromo, 1-benzofuran-3-yl-2-bromo-1-ethanone, pubchem7014, 3-2-bromoacetyl benzo b furan, 1-benzofuran-3-yl-2-bromo-ethanone |
PubChem CID | 2776580 |
IUPAC Name | 1-(1-benzofuran-3-yl)-2-bromoethanone |
SMILES | BrCC(=O)C1=COC2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10681863
|
Thermo Scientific Maybridge
CC27505CB |
250 mg |
£24.20
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10394612
|
Thermo Scientific Maybridge
CC27505DA |
1 g |
£72.55
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10306764
|
Thermo Scientific Maybridge
CC27505DE |
5 g |
£200.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
187657-92-7 | |
239.07 | |
WVHFTONHSRLBGL-UHFFFAOYSA-N | |
2776580 | |
BrCC(=O)C1=COC2=CC=CC=C12 |
C10H7BrO2 | |
MFCD03407369 | |
1-benzofuran-3-yl-2-bromoethanone, 1-1-benzofuran-3-yl-2-bromo-1-ethanone, 1-1-benzofuran-3-yl-2-bromoethanone, 1-benzofuran-3-yl-2-bromoethan-1-one, 1-1-benzofuran-3-yl-2-bromoethan-1-one, ethanone,1-3-benzofuranyl-2-bromo, 1-benzofuran-3-yl-2-bromo-1-ethanone, pubchem7014, 3-2-bromoacetyl benzo b furan, 1-benzofuran-3-yl-2-bromo-ethanone | |
1-(1-benzofuran-3-yl)-2-bromoethanone |
Specifications
187657-92-7 | |
C10H7BrO2 | |
1-benzofuran-3-yl-2-bromoethanone, 1-1-benzofuran-3-yl-2-bromo-1-ethanone, 1-1-benzofuran-3-yl-2-bromoethanone, 1-benzofuran-3-yl-2-bromoethan-1-one, 1-1-benzofuran-3-yl-2-bromoethan-1-one, ethanone,1-3-benzofuranyl-2-bromo, 1-benzofuran-3-yl-2-bromo-1-ethanone, pubchem7014, 3-2-bromoacetyl benzo b furan, 1-benzofuran-3-yl-2-bromo-ethanone | |
BrCC(=O)C1=COC2=CC=CC=C12 | |
239.07 | |
239.07 | |
1-(1-Benzofuran-3-yl)-2-bromo-1-ethanone |
95% | |
MFCD03407369 | |
WVHFTONHSRLBGL-UHFFFAOYSA-N | |
1-(1-benzofuran-3-yl)-2-bromoethanone | |
2776580 | |
95% |