Learn More
1,1'-Carbonyldiimidazole, 97%, Thermo Scientific Chemicals
£27.00 - £676.00
Chemical Identifiers
CAS | 530-62-1 |
---|---|
Molecular Formula | C7H6N4O |
Molecular Weight (g/mol) | 162.15 |
MDL Number | MFCD00005286 |
InChI Key | PFKFTWBEEFSNDU-UHFFFAOYSA-N |
Synonym | 1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole |
PubChem CID | 68263 |
IUPAC Name | di(imidazol-1-yl)methanone |
SMILES | O=C(N1C=CN=C1)N1C=CN=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10675392
|
Thermo Scientific Acros
151810100 |
10 g | Glass bottle | N/A | |||||
10123010
|
Thermo Scientific Acros
151810250 |
25 g | Glass Bottle | N/A | |||||
10705821
|
Thermo Scientific Acros
151811000 |
100 g | Glass bottle | N/A | |||||
10553674
|
Thermo Scientific Acros
151815000 |
500 g | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
530-62-1 | |
162.15 | |
PFKFTWBEEFSNDU-UHFFFAOYSA-N | |
68263 | |
O=C(N1C=CN=C1)N1C=CN=C1 |
C7H6N4O | |
MFCD00005286 | |
1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole | |
di(imidazol-1-yl)methanone |
Specifications
530-62-1 | |
100.0 | |
White to Cream | |
96% min | |
MFCD00005286 | |
01,114; 02,61; 05,97; 06,97; 08,77; 09,96; 12,106; 13,66; 16,64 | |
1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole | |
PFKFTWBEEFSNDU-UHFFFAOYSA-N | |
di(imidazol-1-yl)methanone | |
68263 | |
97% | |
1,1′-Carbonyldiimidazole |
96.0 | |
118.0°C to 120.0°C | |
Authentic | |
C7H6N4O | |
23, V,4, 245 | |
15, 1820 | |
Solubility in water: decomposes. Other solubilities: moderately soluble in thf, soluble in ethanol | |
O=C(N1C=CN=C1)N1C=CN=C1 | |
162.15 | |
162.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-488-9
- 1,1``-Carbonyldiimidazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Reproductive toxicity Category 1B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H360D-May damage the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only