Learn More
1,1'-Carbonyldiimidazole, 97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 118.0°C to 120.0°C |
Color | White to Cream |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min |
Packaging | Glass bottle |
Quantity | 100 g |
Beilstein | 23, V,4, 245 |
Fieser | 01,114; 02,61; 05,97; 06,97; 08,77; 09,96; 12,106; 13,66; 16,64 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464386 | Certificate of Analysis | 26/07/2024 | 151810250,151815000,151811000,151810100 |
A0461553 | Certificate of Analysis | 08/07/2024 | 151810250,151815000,151811000,151810100 |
A0457812 | Certificate of Analysis | 05/01/2024 | 151810250,151815000,151811000,151810100 |
A0453442 | Certificate of Analysis | 25/08/2023 | 151810250,151815000,151811000,151810100 |
A0395051 | Certificate of Analysis | 15/12/2022 | 151810250,151815000,151811000,151810100 |
Safety and Handling



- 1,1``-Carbonyldiimidazole
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Reproductive toxicity Category 1B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H360D-May damage the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.