Learn More
1,10-Diaza-18-crown-6, 96%, Thermo Scientific Chemicals
£44.52 - £371.85
Chemical Identifiers
CAS | 23978-55-4 |
---|---|
Molecular Formula | C12H28N2O4 |
Molecular Weight (g/mol) | 264.37 |
MDL Number | MFCD00005112 |
InChI Key | NLMDJJTUQPXZFG-UHFFFAOYSA-P |
Synonym | kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane |
PubChem CID | 72805 |
IUPAC Name | 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane |
SMILES | C1COCCOCC[NH2+]CCOCCOCC[NH2+]1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15427947
|
Thermo Scientific Alfa Aesar
H51063.MD |
250 mg |
£44.52
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407947
|
Thermo Scientific Alfa Aesar
H51063.03 |
1 g |
£106.53
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15417947
|
Thermo Scientific Alfa Aesar
H51063.06 |
5 g |
£371.85
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
1,10-Diaza-18-crown-6 are starting materials to produce macrocyclic polyamine metal-complexing agents. It is used as a ligand in chemistry for the applications of diagnostic imaging technique to identify medical condition or disease and for the development of medication as a therapeutic agent and pharmaceutic aid. Also used as an ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemische Identifikatoren
23978-55-4 | |
264.37 | |
NLMDJJTUQPXZFG-UHFFFAOYSA-P | |
72805 | |
C1COCCOCC[NH2+]CCOCCOCC[NH2+]1 |
C12H28N2O4 | |
MFCD00005112 | |
kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane | |
1,4,10,13-tetraoxa-7,16-diazacyclooctadecane |
Spezifikation
23978-55-4 | |
MFCD00005112 | |
Air Sensitive | |
Soluble in water. | |
C1COCCOCC[NH2+]CCOCCOCC[NH2+]1 | |
264.37 | |
262.35 | |
1,10-Diaza-18-crown-6 |
C12H28N2O4 | |
609764 | |
kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane | |
NLMDJJTUQPXZFG-UHFFFAOYSA-P | |
1,4,10,13-tetraoxa-7,16-diazacyclooctadecane | |
72805 | |
96% |
Sicherheit und Handhabung
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 245-965-0
missing translation for 'rtecsNumber' : HM1650000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.