Learn More
1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals
Oxidation, reduction indicator
£70.80 - £227.00
Chemical Identifiers
CAS | 14586-54-0 |
---|---|
Molecular Formula | C36H24Cl2FeN6O8 |
Molecular Weight (g/mol) | 795.367 |
MDL Number | MFCD00050959 |
InChI Key | QIZDTKUCQUZJHS-UHFFFAOYSA-L |
Synonym | 1,10-phenanthroline ferrous perchlorate, iron 2+ ; 1,10-phenanthroline; diperchlorate, iron 2+ perchlorate 1,10-phenanthroline 1:2:3, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, diperchlorate, tris 1,10-phenanthroline-n1,n10 iron diperchlorate, 3c12h8n2.2clo4.fe, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, diperchlorate, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, perchlorate 1:2, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, perchlorate 1:2 |
PubChem CID | 84537 |
IUPAC Name | iron(2+);1,10-phenanthroline;diperchlorate |
SMILES | C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Fe+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357308
|
Thermo Scientific Alfa Aesar
030557.04 |
2 g | N/A | ||||||
11367308
|
Thermo Scientific Alfa Aesar
030557.09 |
10 g |
£227.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,10-Phenanthroline iron(II) perchlorate is used as an oxidation, reduction indicator. It is used for the detection, identification, analysis, of chemical, biological, or pathologic processes or conditions. Agents that are capable of inserting themselves between the successive bases in DNA, thus kinking, uncoiling or otherwise deforming it and therefore preventing its proper functioning. They are used in the study of DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14586-54-0 | |
795.367 | |
QIZDTKUCQUZJHS-UHFFFAOYSA-L | |
84537 | |
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Fe+2] |
C36H24Cl2FeN6O8 | |
MFCD00050959 | |
1,10-phenanthroline ferrous perchlorate, iron 2+ ; 1,10-phenanthroline; diperchlorate, iron 2+ perchlorate 1,10-phenanthroline 1:2:3, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, diperchlorate, tris 1,10-phenanthroline-n1,n10 iron diperchlorate, 3c12h8n2.2clo4.fe, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, diperchlorate, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, perchlorate 1:2, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, perchlorate 1:2 | |
iron(2+);1,10-phenanthroline;diperchlorate |
Specifications
14586-54-0 | |
C36H24Cl2FeN6O8 | |
UN1479 | |
QIZDTKUCQUZJHS-UHFFFAOYSA-L | |
iron(2+);1,10-phenanthroline;diperchlorate | |
84537 | |
Crystalline |
Red | |
MFCD00050959 | |
1,10-phenanthroline ferrous perchlorate, iron 2+ ; 1,10-phenanthroline; diperchlorate, iron 2+ perchlorate 1,10-phenanthroline 1:2:3, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, diperchlorate, tris 1,10-phenanthroline-n1,n10 iron diperchlorate, 3c12h8n2.2clo4.fe, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, diperchlorate, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, perchlorate 1:2, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, perchlorate 1:2 | |
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Fe+2] | |
795.367 | |
795.37 | |
1,10-Phenanthroline iron(II) perchlorate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
missing translation for 'tsca' : TSCA
- 1,10-Phenanthroline iron(II) perchlorate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.