missing translation for 'onlineSavingsMsg'
Learn More
1,2,3,6-Tetrahydropyridine, 98%, Thermo Scientific Chemicals Product Code.: 10585134

1,2,3,6-Tetrahydropyridine, 98%, Thermo Scientific Chemicals

Product Code. 10585134
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10585134

Brand: Thermo Scientific Acros 158870010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-05-3
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00005949
InChI Key FTAHXMZRJCZXDL-UHFFFAOYSA-N
Synonym pyridine, 1,2,3,6-tetrahydro, 3-piperideine, 1,2,5,6-tetrahydropyridine, unii-26rls9d255, delta 3-piperideine, .delta.3-piperidine, delta3-piperidine, acmc-1bcyc, 1,3,6-tetrahydropyridine, 1,5,6-tetrahydropyridine
PubChem CID 12750
ChEBI CHEBI:47860
IUPAC Name 1,2,3,6-tetrahydropyridine
SMILES C1CNCC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -48.0°C
Color Cream-Yellow to White
Density 0.9100g/mL
Boiling Point 108.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.48
Quantity 1 g
Beilstein 20, III, 1912
Specific Gravity 0.91
Solubility Information Solubility in water: soluble.
Formula Weight 83.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,6-Tetrahydropyridine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0454209 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number158870010,158870050
Lot NumberA0397871 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number
Lot NumberA0404740 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number
Lot NumberA0441102 Certificate TypeCertificate of Analysis Date21/04/2022 Catalog Number
Lot NumberA0431939 Certificate TypeCertificate of Analysis Date02/07/2021 Catalog Number
compliance-icons compliance-icons
Product Identifier
  • 1,2,3,6-Tetrahydropyridine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2,3,6-Tetrahydropyridine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.