Learn More
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
£14.50 - £156.00
Chemical Identifiers
CAS | 288-88-0 |
---|---|
Molecular Formula | C2H3N3 |
Molecular Weight (g/mol) | 69.06 |
MDL Number | MFCD00005228 |
InChI Key | NSPMIYGKQJPBQR-UHFFFAOYSA-N |
Synonym | 1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van |
PubChem CID | 9257 |
ChEBI | CHEBI:35550 |
IUPAC Name | 1H-1,2,4-triazole |
SMILES | C1=NC=NN1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10081510
|
Thermo Scientific Acros
139280050 |
5 g | Glass Bottle |
£14.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10081520
|
Thermo Scientific Acros
139280250 |
25 g | Plastic bottle |
£20.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10457150
|
Thermo Scientific Acros
139281000 |
100 g | Plastic bottle |
£49.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10532164
|
Thermo Scientific Acros
139285000 |
500 g | Plastic bottle |
£156.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
288-88-0 | |
69.06 | |
NSPMIYGKQJPBQR-UHFFFAOYSA-N | |
9257 | |
1H-1,2,4-triazole |
C2H3N3 | |
MFCD00005228 | |
1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van | |
CHEBI:35550 | |
C1=NC=NN1 |
Specifications
288-88-0 | |
100.0 | |
White | |
140°C | |
99.4% min. (HPLC) | |
MFCD00005228 | |
01,1188; 02,423 | |
1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van | |
NSPMIYGKQJPBQR-UHFFFAOYSA-N | |
1H-1,2,4-triazole | |
9257 | |
69.06 | |
Crystalline Powder or Flakes |
99.4 | |
119°C to 122°C | |
260°C | |
Authentic | |
C2H3N3 | |
26, 13 | |
15, 9766 | |
Solubility in water: 1250g/L (20°C). Other solubilities: soluble in chloroform,xylene,ethylacetate and,alcohol | |
C1=NC=NN1 | |
69.06 | |
CHEBI:35550 | |
99.50% | |
1, 2, 4-1H-Triazole |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Suspected of damaging the unborn child.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 206-022-9
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only