missing translation for 'onlineSavingsMsg'
Learn More
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals Product Code.: 10081520

1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals

Product Code. 10081520
25 g, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10081520

Brand: Thermo Scientific Acros 139280250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-88-0
Molecular Formula C2H3N3
Molecular Weight (g/mol) 69.06
MDL Number MFCD00005228
InChI Key NSPMIYGKQJPBQR-UHFFFAOYSA-N
Synonym 1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole vanShow MoreShow Less
PubChem CID 9257
ChEBI CHEBI:35550
IUPAC Name 1H-1,2,4-triazole
SMILES C1=NC=NN1

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point 119°C to 122°C
Color White
Boiling Point 260°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (HPLC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 26, 13
Fieser 01,1188; 02,423
Merck Index 15, 9766
Solubility Information Solubility in water: 1250g/L (20°C). Other solubilities: soluble in chloroform,xylene,ethylacetate and,alcohol
Formula Weight 69.06
Percent Purity 99.50%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 1, 2, 4-1H-Triazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468667 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number139285000,139281000,139280250
Lot NumberA0468914 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number139285000,139281000,139280250
Lot NumberA0432869 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number139285000,139281000,139280250,139280050
Lot NumberA0215282 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number139285000,139281000,139280250,139280050
Lot NumberA0230514 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number139285000,139281000,139280250,139280050
compliance-icons compliance-icons
Product Identifier
  • 1,2,4-1H-Triazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.