missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals

£25.33 - £84.52

Chemical Identifiers

CAS 95-93-2
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
InChI Key SQNZJJAZBFDUTD-UHFFFAOYSA-N
Synonym durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren
PubChem CID 7269
ChEBI CHEBI:38978
IUPAC Name 1,2,4,5-tetramethylbenzene
SMILES CC1=CC(=C(C=C1C)C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15547029
View Documents
Thermo Scientific Acros
138370050
5 g
£25.33
5g
Please to purchase this item. Need a web account? Register with us today!
15502245
View Documents Promotion Details
Thermo Scientific Acros
138371000
100 g
£25.93
100g
Please to purchase this item. Need a web account? Register with us today!
15557029
View Documents
Thermo Scientific Acros
138375000
500 g
£84.52
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

95-93-2
134.22
durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren
CHEBI:38978
CC1=CC(=C(C=C1C)C)C
C10H14
SQNZJJAZBFDUTD-UHFFFAOYSA-N
7269
1,2,4,5-tetramethylbenzene

Specifications

95-93-2
196.0°C to 197.0°C
Authentic
Glass bottle
C6H2(CH3)4
15,3515
Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene
CC1=CC(=C(C=C1C)C)C
134.22
CHEBI:38978
97+%
78.0°C to 82.0°C
73°C
97% min. (GC)
C10H14
05,431
durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren
SQNZJJAZBFDUTD-UHFFFAOYSA-N
1,2,4,5-tetramethylbenzene
7269
134.22
1,2,4,5-Tetramethylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-465-7

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only