missing translation for 'onlineSavingsMsg'
Learn More
1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals Product Code.: 15431017

1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals

Product Code. 15431017
10 kg
Click to view available options
Quantity:
500 g
2 kg
10 kg
This item is not returnable. View return policy

Product Code. 15431017

Brand: Thermo Scientific Alfa Aesar 012411.A9

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,2,4-Trimethylbenzene is applied as a liquid scintillator. It is also used as a sterilizing agent and in the manufacture of dyes, perfumes, and resins. It is used as a safe solvent for the production of H2O2 (hydrogen peroxide). Pseudocumene is used mainly as raw material for the production of trimellitic anhydride, pyromellitic dianhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-63-6
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008527
InChI Key GWHJZXXIDMPWGX-UHFFFAOYSA-N
Synonym pseudocumene, pseudocumol, psi-cumene, as-trimethylbenzene, 1,3,4-trimethylbenzene, benzene, 1,2,4-trimethyl, uns-trimethylbenzene, 1,2,5-trimethylbenzene, asymmetrical trimethylbenzene, .psi.-cumeneShow MoreShow Less
PubChem CID 7247
ChEBI CHEBI:34039
IUPAC Name 1,2,4-trimethylbenzene
SMILES CC1=CC(=C(C=C1)C)C

Specifications

Melting Point -44°C
Density 0.882 g/mL
Boiling Point 169°C to 171°C
Flash Point 48°C (118°F)
Odor Aromatic
Refractive Index 1.504
Quantity 10 kg
UN Number UN3295
Beilstein 1903005
Merck Index 14,7915
Solubility Information Soluble in alcohol,benzene and ether; Slightly soluble in water
Formula Weight 120.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,2,4-Trimethylbenzene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW05K032 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number012411.36,012411.A3,012411.B4,012411.A9
Lot NumberM31K023 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog Number012411.36,012411.A3,012411.B4,012411.A9
Lot NumberM11I017 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012411.36,012411.A3,012411.B4,012411.A9
Lot NumberZ12A003 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012411.36,012411.A3,012411.B4,012411.A9
Lot NumberC12P38 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012411.36,012411.A3,012411.B4,012411.A9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.