missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific™
POA - £210.00
Chemical Identifiers
CAS | 569650-64-2 |
---|---|
Molecular Formula | C34H56BF4P2Rh |
Molecular Weight (g/mol) | 716.48 |
MDL Number | MFCD07369039 |
InChI Key | AXLKBYDAACXFBH-KFORCWLUNA-N |
Synonym | 1,2-bis 2r,5r-2,5-diisopropylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate, r,r-i-pr-duphos-rh, +-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate |
PubChem CID | 12964416 |
IUPAC Name | λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-bis(propan-2-yl)phospholan-1-yl]phenyl}-2,5-bis(propan-2-yl)phospholane tetrafluoroboranuide |
SMILES | [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@@H]1CC[C@@H](C(C)C)P1C1=CC=CC=C1P1[C@@H](CC[C@H]1C(C)C)C(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10047999
|
Thermo Scientific Acros
435200500 |
50 mg | |||||||
10331150
|
Thermo Scientific Acros
435202500 |
250 mg |
£210.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
569650-64-2 | |
716.48 | |
AXLKBYDAACXFBH-KFORCWLUNA-N | |
12964416 | |
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@@H]1CC[C@@H](C(C)C)P1C1=CC=CC=C1P1[C@@H](CC[C@H]1C(C)C)C(C)C |
C34H56BF4P2Rh | |
MFCD07369039 | |
1,2-bis 2r,5r-2,5-diisopropylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate, r,r-i-pr-duphos-rh, +-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate | |
λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-bis(propan-2-yl)phospholan-1-yl]phenyl}-2,5-bis(propan-2-yl)phospholane tetrafluoroboranuide |
Specifications
569650-64-2 | |
100.0 | |
54.7 to 59.3% C | |
97% | |
MFCD07369039 | |
AXLKBYDAACXFBH-KFORCWLUNA-N | |
λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-bis(propan-2-yl)phospholan-1-yl]phenyl}-2,5-bis(propan-2-yl)phospholane tetrafluoroboranuide | |
12964416 | |
97% | |
1,2-Bis((2R,5R)-2,5-Diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate |
96.0 | |
Brown to Yellow | |
Authentic | |
C34H56BF4P2Rh | |
1,2-bis 2r,5r-2,5-diisopropylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate, r,r-i-pr-duphos-rh, +-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate | |
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@@H]1CC[C@@H](C(C)C)P1C1=CC=CC=C1P1[C@@H](CC[C@H]1C(C)C)C(C)C | |
716.48 | |
716.48 | |
Powder |
RUO – Research Use Only