missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[2-(Boc-amino)ethyl]indole-6-carboxylic acid, 97%, Thermo Scientific™
£24.40 - £472.00
Chemical Identifiers
CAS | 885266-67-1 |
---|---|
Molecular Formula | C9H5N3O2S |
Molecular Weight (g/mol) | 219.218 |
MDL Number | MFCD04973974 |
InChI Key | FDQWHYSXVRDVMV-UHFFFAOYSA-N |
Synonym | 4-nitro-1h-indol-3-yl sulfanyl formonitrile |
PubChem CID | 24720942 |
IUPAC Name | (4-nitro-1H-indol-3-yl) thiocyanate |
SMILES | C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)SC#N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
885266-67-1 | |
219.218 | |
FDQWHYSXVRDVMV-UHFFFAOYSA-N | |
24720942 | |
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)SC#N |
C9H5N3O2S | |
MFCD04973974 | |
4-nitro-1h-indol-3-yl sulfanyl formonitrile | |
(4-nitro-1H-indol-3-yl) thiocyanate |
Specifications
885266-67-1 | |
C9H5N3O2S | |
4-nitro-1h-indol-3-yl sulfanyl formonitrile | |
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)SC#N | |
219.218 | |
304.34 | |
Solid |
White | |
MFCD04973974 | |
FDQWHYSXVRDVMV-UHFFFAOYSA-N | |
(4-nitro-1H-indol-3-yl) thiocyanate | |
24720942 | |
97% | |
1-[2-(Boc-amino)ethyl]indole-6-carboxylic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold