missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-2,4-dichlorobenzene, 97+%, Thermo Scientific™

£17.78 - £247.00

Chemical Identifiers

CAS 6954-77-4
Molecular Formula C8H7BrCl2O
Molecular Weight (g/mol) 269.95
MDL Number MFCD00043464
InChI Key OEYZGSRLAGSENP-UHFFFAOYSA-N
Synonym 1-2-bromoethoxy-2,4-dichlorobenzene, beta-bromo-2,4-dichlorophenetole, b-bromo-2,4-dichlorophenetole, 1-2,4-dichlorophenoxy-2-bromoethane, 1-bromo-2-2,4-dichlorophenoxy ethane, pubchem3151, bromoethoxydichlorobenzene, 2,4-dichlorophenoxyethylbromide, 2,4-dichlorophenoxyethyl bromide, 2-2,4-dichlorophenoxy ethyl bromide
PubChem CID 138881
IUPAC Name 1-(2-bromoethoxy)-2,4-dichlorobenzene
SMILES ClC1=CC=C(OCCBr)C(Cl)=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15479047
View Documents
Thermo Scientific Alfa Aesar
H52367.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15459047
View Documents
Thermo Scientific Alfa Aesar
H52367.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15469047
View Documents
Thermo Scientific Alfa Aesar
H52367.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6954-77-4
269.95
OEYZGSRLAGSENP-UHFFFAOYSA-N
138881
ClC1=CC=C(OCCBr)C(Cl)=C1
C8H7BrCl2O
MFCD00043464
1-2-bromoethoxy-2,4-dichlorobenzene, beta-bromo-2,4-dichlorophenetole, b-bromo-2,4-dichlorophenetole, 1-2,4-dichlorophenoxy-2-bromoethane, 1-bromo-2-2,4-dichlorophenoxy ethane, pubchem3151, bromoethoxydichlorobenzene, 2,4-dichlorophenoxyethylbromide, 2,4-dichlorophenoxyethyl bromide, 2-2,4-dichlorophenoxy ethyl bromide
1-(2-bromoethoxy)-2,4-dichlorobenzene

Specifications

6954-77-4
MFCD00043464
1-2-bromoethoxy-2,4-dichlorobenzene, beta-bromo-2,4-dichlorophenetole, b-bromo-2,4-dichlorophenetole, 1-2,4-dichlorophenoxy-2-bromoethane, 1-bromo-2-2,4-dichlorophenoxy ethane, pubchem3151, bromoethoxydichlorobenzene, 2,4-dichlorophenoxyethylbromide, 2,4-dichlorophenoxyethyl bromide, 2-2,4-dichlorophenoxy ethyl bromide
ClC1=CC=C(OCCBr)C(Cl)=C1
269.95
269.95
1-(2-Bromoethoxy)-2,4-dichlorobenzene
C8H7BrCl2O
UN2810
OEYZGSRLAGSENP-UHFFFAOYSA-N
1-(2-bromoethoxy)-2,4-dichlorobenzene
138881
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications