missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-1-iodotrifluoroethane, 97%, Thermo Scientific™

£9.30 - £22.00

Chemical Identifiers

CAS 216394-01-3
Molecular Formula C2Br2F3I
Molecular Weight (g/mol) 367.73
MDL Number MFCD00042521
InChI Key YUBJHLYGOOZDRN-UHFFFAOYNA-N
Synonym 1,2-dibromo-1-iodotrifluoroethane, 1,2-dibromo-2-iodotrifluoroethane, 1,2-dibromo-1,2,2-trifluoro-1-iodoethane, ethane,1,2-dibromo-1,1,2-trifluoro-2-iodo
PubChem CID 2736780
IUPAC Name 1,2-dibromo-1,1,2-trifluoro-2-iodoethane
SMILES FC(F)(Br)C(F)(Br)I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11437104
View Documents
Thermo Scientific Alfa Aesar
B21532.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11447104
View Documents
Thermo Scientific Alfa Aesar
B21532.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

216394-01-3
367.73
YUBJHLYGOOZDRN-UHFFFAOYNA-N
2736780
FC(F)(Br)C(F)(Br)I
C2Br2F3I
MFCD00042521
1,2-dibromo-1-iodotrifluoroethane, 1,2-dibromo-2-iodotrifluoroethane, 1,2-dibromo-1,2,2-trifluoro-1-iodoethane, ethane,1,2-dibromo-1,1,2-trifluoro-2-iodo
1,2-dibromo-1,1,2-trifluoro-2-iodoethane

Specifications

216394-01-3
63°C to 65°C (32 mmHg)
BrCFICF2Br
1,2-dibromo-1-iodotrifluoroethane, 1,2-dibromo-2-iodotrifluoroethane, 1,2-dibromo-1,2,2-trifluoro-1-iodoethane, ethane,1,2-dibromo-1,1,2-trifluoro-2-iodo
FC(F)(Br)C(F)(Br)I
367.73
367.73
1,2-Dibromo-1-iodotrifluoroethane
1.515
C2Br2F3I
MFCD00042521
YUBJHLYGOOZDRN-UHFFFAOYNA-N
1,2-dibromo-1,1,2-trifluoro-2-iodoethane
2736780
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications