missing translation for 'onlineSavingsMsg'
Learn More
1,2-Dichlorobenzene, 99%, pure, Thermo Scientific Chemicals Product Code.: 10530371

1,2-Dichlorobenzene, 99%, pure, Thermo Scientific Chemicals

Product Code. 10530371
1 L, Glass Bottle
Click to view available options
Quantity:
5 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
This item is not returnable. View return policy

Product Code. 10530371

Brand: Thermo Scientific Acros 113180010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-50-1
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, clorobenShow MoreShow Less
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -15.0°C
Color Colorless to Yellow
Density 1.3000g/mL
Boiling Point 179.0°C to 180.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H4Cl2
Linear Formula C6H4Cl2
Refractive Index 1.5500 to 1.552
Quantity 1 L
UN Number 1591
Beilstein 05, 201
Merck Index 15, 3065
Specific Gravity 1.3
Solubility Information Solubility in water: 0.13g/L(20°C). Other solubilities: miscible with most common organic solvents,miscible with alcohol,ether and benzene
Molecular Weight (g/mol) 147
Formula Weight 147
Vapor Pressure 1.3mbar at 20°C
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1, 2-Dichlorobenzene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467084 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog Number113180010,113180025,113180250
Lot NumberA0462071 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog Number113180010,113180025,113180050,113180250
Lot NumberB0541082A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number113180010,113180025,113180050,113180250
Lot NumberB0540470A Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number113180010,113180025,113180050,113180250
Lot NumberB0539169A Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number113180010,113180025,113180050,113180250
compliance-icons compliance-icons
Product Identifier
  • 1,2-Dichlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Dichlorobenzene, 99%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.