Learn More
1,2-Dichloroethane, 99.5%, Thermo Scientific Chemicals
£18.35 - £85.80
Chemical Identifiers
CAS | 107-06-2 |
---|---|
Molecular Formula | C2H4Cl2 |
Molecular Weight (g/mol) | 98.95 |
MDL Number | MFCD00000963 |
InChI Key | WSLDOOZREJYCGB-UHFFFAOYSA-N |
Synonym | ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion |
PubChem CID | 11 |
ChEBI | CHEBI:27789 |
IUPAC Name | 1,2-dichloroethane |
SMILES | ClCCCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15352032
|
Thermo Scientific Acros
443011000 |
100 mL |
£18.35
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15342032
|
Thermo Scientific Acros
443010010 |
1 L |
£66.70
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11934471
|
Thermo Scientific Acros
443010025 |
2.5 L | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-06-2 | |
98.95 | |
WSLDOOZREJYCGB-UHFFFAOYSA-N | |
11 | |
1,2-dichloroethane |
C2H4Cl2 | |
MFCD00000963 | |
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion | |
CHEBI:27789 | |
ClCCCl |
Specifications
-35.0°C | |
1.2500g/mL | |
13°C | |
1.25 | |
ClCH2CH2Cl | |
0.8 mPa.s (20°C) | |
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion | |
WSLDOOZREJYCGB-UHFFFAOYSA-N | |
1,2-dichloroethane | |
11 | |
98.96 | |
Extra Dry over Molecular Sieve |
107-06-2 | |
81.0°C to 85.0°C | |
1.4440 to 1.4460 | |
C2H4Cl2 | |
MFCD00000963 | |
0.005% max. | |
Solubility in water: 8.7g/L (20°C). Other solubilities: miscible with most common organic solvents | |
ClCCCl | |
98.95 | |
CHEBI:27789 | |
99.5% | |
1,2-Dichloroethane |
Safety and Handling
GHS H Statement May cause cancer. Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. Highly flammable liquid and vapor. Harmful if swallowed. Toxic if inhaled. May cause drowsiness or dizziness.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger
- 1,2-Dichloroethane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 3
- Specific target organ toxicity Category 3
- Carcinogenicity Category 1B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only