missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Honeywell™
Reagent Grade, inhibitor-free,≥99%
£29.90 - £1636.14
Chemical Identifiers
CAS | 110-71-4 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.122 |
MDL Number | MFCD00008502 |
InChI Key | XTHFKEDIFFGKHM-UHFFFAOYSA-N |
Synonym | monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl etherShow More |
PubChem CID | 8071 |
ChEBI | CHEBI:42263 |
IUPAC Name | 1,2-dimethoxyethane |
SMILES | COCCOC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
15647840
|
Honeywell Research Chemicals
E27408-100ML |
100 mL | Glass bottle | N/A | ||||||
15687840
|
Honeywell Research Chemicals
E27408-500ML |
500 mL | Glass bottle | N/A | ||||||
15657840
|
Honeywell Research Chemicals
E27408-1L |
1 L | Glass bottle |
£106.86
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15667840
|
Honeywell Research Chemicals
E27408-2.5L |
2.5 L | Glass bottle |
£258.61
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15677840
|
Honeywell Research Chemicals
E27408-20L |
20 L | Stainless steel can |
£1636.14
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
110-71-4 | |
90.122 | |
XTHFKEDIFFGKHM-UHFFFAOYSA-N | |
8071 | |
1,2-dimethoxyethane |
C4H10O2 | |
MFCD00008502 | |
monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl etherShow More | |
CHEBI:42263 | |
COCCOC |
Specifications
110-71-4 | |
Colorless | |
Neutral | |
−2°C (28.4°F) | |
CH3OCH2CH2OCH3 | |
UN2252 | |
monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether | |
COCCOC | |
90.122 | |
3.1 (20°C, vs air) | |
90.12g/mol | |
1,2-Dimethoxyethane |
-58°C | |
0.867g/mL at 25°C | |
85°C (1013 hPa) | |
C4H10O2 | |
MFCD00008502 | |
1209237 | |
XTHFKEDIFFGKHM-UHFFFAOYSA-N | |
1,2-dimethoxyethane | |
8071 | |
CHEBI:42263 | |
≥99% |
Safety and Handling
P201-P210-P280-P308 + P313-P370 + P378-P403 + P235
missing translation for 'dotInformation' : 3/PGII
missing translation for 'rtecsNumber' : 203-794-9
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Honeywell™