missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Fluorophenyl)pyrrole, 98%, Thermo Scientific™
£19.10 - £150.00
Chemical Identifiers
CAS | 89096-77-5 |
---|---|
Molecular Formula | C10H8FN |
Molecular Weight (g/mol) | 161.18 |
MDL Number | MFCD02665231 |
InChI Key | LMLYMAWSPZKOFR-UHFFFAOYSA-N |
Synonym | 1-2-fluorophenyl-1h-pyrrole, 1-2-fluorophenyl pyrrole, 1h-pyrrole, 1-2-fluorophenyl, n-2-fluorophenyl pyrrole, acmc-20lhmh, 1-2-fluoro-phenyl-1h-pyrrole |
PubChem CID | 2767601 |
SMILES | FC1=CC=CC=C1N1C=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15406067
|
Thermo Scientific Alfa Aesar
H33247.03 |
1 g |
£19.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15416067
|
Thermo Scientific Alfa Aesar
H33247.06 |
5 g | N/A | ||||||
15426067
|
Thermo Scientific Alfa Aesar
H33247.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
89096-77-5 | |
161.18 | |
LMLYMAWSPZKOFR-UHFFFAOYSA-N | |
2767601 |
C10H8FN | |
MFCD02665231 | |
1-2-fluorophenyl-1h-pyrrole, 1-2-fluorophenyl pyrrole, 1h-pyrrole, 1-2-fluorophenyl, n-2-fluorophenyl pyrrole, acmc-20lhmh, 1-2-fluoro-phenyl-1h-pyrrole | |
FC1=CC=CC=C1N1C=CC=C1 |
Specifications
89096-77-5 | |
C10H8FN | |
MFCD02665231 | |
LMLYMAWSPZKOFR-UHFFFAOYSA-N | |
1-(2-fluorophenyl)pyrrole | |
2767601 | |
98% |
64°C (1.1 mmHg) | |
1.5105 | |
1-2-fluorophenyl-1h-pyrrole, 1-2-fluorophenyl pyrrole, 1h-pyrrole, 1-2-fluorophenyl, n-2-fluorophenyl pyrrole, acmc-20lhmh, 1-2-fluoro-phenyl-1h-pyrrole | |
FC1=CC=CC=C1N1C=CC=C1 | |
161.18 | |
161.18 | |
1-(2-Fluorophenyl)pyrrole |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title