missing translation for 'onlineSavingsMsg'
Learn More
1,2-Propanediol, 99%, extra pure, Thermo Scientific Chemicals Product Code.: 10377950

1,2-Propanediol, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10377950
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
This item is not returnable. View return policy

Product Code. 10377950

Brand: Thermo Scientific Acros 158720010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.09
MDL Number MFCD00064272
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrostShow MoreShow Less
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -60.0°C
Density 1.0300g/mL
Boiling Point 187.0°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH2OH
Refractive Index 1.4310 to 1.4330
Quantity 1 L
Beilstein 01, 472
Merck Index 15, 7968
Specific Gravity 1.03
Solubility Information Solubility in water: completely soluble. Other solubilities: soluble in ether, miscible with acetone and chloroform
Viscosity 45 mPa.s (20°C)
Formula Weight 76.09
Percent Purity 99%
Grade Extra Pure
Physical Form Viscous Liquid
Chemical Name or Material 1, 2-Propanediol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467983 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number158720025,158720010,158720250
Lot NumberA0461778 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number158720025,158720010,158720250
Lot NumberA0451881 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number158720025,158720010,158720250
Lot NumberA0449838 Certificate TypeCertificate of Analysis Date03/02/2023 Catalog Number158720025,158720010,158720250
Lot NumberA0399689 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number158720025,158720010,158720250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Propanediol, 99%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.