missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Propanediol, 99%, extra pure, Thermo Scientific Chemicals
£26.50 - £348.00
Chemical Identifiers
CAS | 57-55-6 |
---|---|
Molecular Formula | C3H8O2 |
Molecular Weight (g/mol) | 76.09 |
MDL Number | MFCD00064272 |
InChI Key | DNIAPMSPPWPWGF-UHFFFAOYSA-N |
Synonym | 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost |
PubChem CID | 1030 |
ChEBI | CHEBI:16997 |
IUPAC Name | propane-1,2-diol |
SMILES | CC(CO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10377950
|
Thermo Scientific Acros
158720010 |
1 L | Glass Bottle |
£26.50
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10542941
|
Thermo Scientific Acros
158720025 |
2.5 L | Glass Bottle |
£50.70
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10771544
|
Thermo Scientific
158720250 |
25 L | Metal drum |
£348.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57-55-6 | |
76.09 | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
1030 | |
propane-1,2-diol |
C3H8O2 | |
MFCD00064272 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
CHEBI:16997 | |
CC(CO)O |
Specifications
57-55-6 | |
100.0 | |
1.0300g/mL | |
99°C | |
98.5% min. (GC) | |
CH3CH(OH)CH2OH | |
MFCD00064272 | |
15, 7968 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
propane-1,2-diol | |
1030 | |
45 mPa.s (20°C) | |
99% | |
Viscous Liquid |
98.5 | |
-60.0°C | |
187.0°C | |
Authentic | |
C3H8O2 | |
1.4310 to 1.4330 | |
01, 472 | |
1.03 | |
Solubility in water: completely soluble. Other solubilities: soluble in ether, miscible with acetone and chloroform | |
CC(CO)O | |
76.09 | |
CHEBI:16997 | |
76.09 | |
Extra Pure | |
1, 2-Propanediol |
Safety and Handling
missing translation for 'einecsNumber' : 200-338-
RUO – Research Use Only