missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one, 97%, Thermo Scientific™
£12.75 - £174.00
Chemical Identifiers
CAS | 175137-02-7 |
---|---|
Molecular Formula | C12H10ClNOS |
Molecular Weight (g/mol) | 251.73 |
MDL Number | MFCD00068162 |
InChI Key | DLQVOIFYJQATMI-UHFFFAOYSA-N |
Synonym | 1-3-amino-5-4-chlorophenyl-2-thienyl ethan-1-one, 1-3-amino-5-4-chlorophenyl thiophen-2-yl ethanone, 2-acetyl-3-amino-5-4-chlorophenyl thiophene, ethanone,1-3-amino-5-4-chlorophenyl-2-thienyl, 1-3-amino-5-4-chlorophenyl thiophen-2-yl ethan-1-one |
PubChem CID | 2777494 |
SMILES | CC(=O)C1=C(N)C=C(S1)C1=CC=C(Cl)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10089251
|
Thermo Scientific Maybridge
GK00604CB |
250 mg |
£12.75
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10456082
|
Thermo Scientific Maybridge
GK00604DA |
1 g |
£33.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10643455
|
Thermo Scientific Maybridge
GK00604EA |
10 g |
£174.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
175137-02-7 | |
251.73 | |
DLQVOIFYJQATMI-UHFFFAOYSA-N | |
2777494 |
C12H10ClNOS | |
MFCD00068162 | |
1-3-amino-5-4-chlorophenyl-2-thienyl ethan-1-one, 1-3-amino-5-4-chlorophenyl thiophen-2-yl ethanone, 2-acetyl-3-amino-5-4-chlorophenyl thiophene, ethanone,1-3-amino-5-4-chlorophenyl-2-thienyl, 1-3-amino-5-4-chlorophenyl thiophen-2-yl ethan-1-one | |
CC(=O)C1=C(N)C=C(S1)C1=CC=C(Cl)C=C1 |
Specifications
175137-02-7 | |
100.0 | |
Amber glass bottle | |
MFCD00068162 | |
DLQVOIFYJQATMI-UHFFFAOYSA-N | |
1-[3-amino-5-(4-chlorophenyl)thiophen-2-yl]ethan-1-one | |
2777494 | |
97% |
96.5 | |
97% | |
C12H10ClNOS | |
1-3-amino-5-4-chlorophenyl-2-thienyl ethan-1-one, 1-3-amino-5-4-chlorophenyl thiophen-2-yl ethanone, 2-acetyl-3-amino-5-4-chlorophenyl thiophene, ethanone,1-3-amino-5-4-chlorophenyl-2-thienyl, 1-3-amino-5-4-chlorophenyl thiophen-2-yl ethan-1-one | |
CC(=O)C1=C(N)C=C(S1)C1=CC=C(Cl)C=C1 | |
251.73 | |
251.74 | |
1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one |