Learn More
1,3-Benzodioxol-5-ylacetyl chloride, ≥97%, Thermo Scientific™
£29.95 - £302.00
Chemical Identifiers
CAS | 6845-81-4 |
---|---|
Molecular Formula | C9H7ClO3 |
Molecular Weight (g/mol) | 198.60 |
MDL Number | MFCD03424722 |
InChI Key | HGSZGCXMAJSUSC-UHFFFAOYSA-N |
Synonym | benzo 1,3 dioxol-5-yl-acetyl chloride, 1,3-benzodioxol-5-ylacetyl chloride, 2-benzo d 1,3 dioxol-5-yl acetyl chloride, benzo 1,3 dioxol-5-ylacetyl chloride, 2-2h-1,3-benzodioxol-5-yl acetyl chloride, 2-2h-benzo 3,4-d 1,3-dioxolen-5-yl acetyl chloride, 1,3-benzodioxole-5-acetylchloride, 3,4-methylenedioxyphenylacetylchloride, 3,4-methylenedioxyphenylacetyl chloride |
PubChem CID | 2756584 |
SMILES | ClC(=O)CC1=CC2=C(OCO2)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10422844
|
Thermo Scientific Maybridge
CC50802CB |
250 mg |
£29.95
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10245624
|
Thermo Scientific Maybridge
CC50802DA |
1 g |
£89.75
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10255624
|
Thermo Scientific Maybridge
CC50802EA |
10 g |
£302.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
6845-81-4 | |
198.60 | |
HGSZGCXMAJSUSC-UHFFFAOYSA-N | |
2756584 |
C9H7ClO3 | |
MFCD03424722 | |
benzo 1,3 dioxol-5-yl-acetyl chloride, 1,3-benzodioxol-5-ylacetyl chloride, 2-benzo d 1,3 dioxol-5-yl acetyl chloride, benzo 1,3 dioxol-5-ylacetyl chloride, 2-2h-1,3-benzodioxol-5-yl acetyl chloride, 2-2h-benzo 3,4-d 1,3-dioxolen-5-yl acetyl chloride, 1,3-benzodioxole-5-acetylchloride, 3,4-methylenedioxyphenylacetylchloride, 3,4-methylenedioxyphenylacetyl chloride | |
ClC(=O)CC1=CC2=C(OCO2)C=C1 |
Specifications
6845-81-4 | |
100.0 | |
Amber glass bottle | |
MFCD03424722 | |
HGSZGCXMAJSUSC-UHFFFAOYSA-N | |
2-(1,3-benzodioxol-5-yl)acetyl chloride | |
2756584 | |
97% |
96.5 | |
97% | |
C9H7ClO3 | |
benzo 1,3 dioxol-5-yl-acetyl chloride, 1,3-benzodioxol-5-ylacetyl chloride, 2-benzo d 1,3 dioxol-5-yl acetyl chloride, benzo 1,3 dioxol-5-ylacetyl chloride, 2-2h-1,3-benzodioxol-5-yl acetyl chloride, 2-2h-benzo 3,4-d 1,3-dioxolen-5-yl acetyl chloride, 1,3-benzodioxole-5-acetylchloride, 3,4-methylenedioxyphenylacetylchloride, 3,4-methylenedioxyphenylacetyl chloride | |
ClC(=O)CC1=CC2=C(OCO2)C=C1 | |
198.60 | |
198.6 | |
1, 3-Benzodioxol-5-ylacetyl chloride |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger